siRNA Details
| Gene Symbol: KHK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000221.2 | NP_000212.1 | 5, 6 | 1082 | ||
| NM_006488.2 | NP_006479.1 | 5, 6 | 1082 | |||
| XM_005264294.3 | XP_005264351.1 | 6, 7 | 1225 | |||
| XM_005264296.3 | XP_005264353.1 | 5, 6 | 1068 | |||
| XM_005264298.3 | XP_005264355.1 | 3, 4 | 835 | |||
| XM_017004060.1 | XP_016859549.1 | 6, 7 | 1222 | |||
| XM_017004061.1 | XP_016859550.1 | 4, 5 | 952 | |||
| GenBank mRNA | AK130033.1 | - | 5, 6 | 599 | ||
| AK223075.1 | - | 5, 6 | 599 | |||
| BC006233.2 | - | 5, 6 | 1020 | 3946893 | ||
| BX648873.1 | - | 3, 4 | 349 | |||
| CR456801.1 | - | 5, 6 | 565 | |||
| X78677.1 | - | 3, 4 | 361 | |||
| X78678.1 | - | 5, 6 | 573 | |||

KHK
Genomic Map