siRNA Details
| Gene Symbol: KIR2DL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080770.1 | NP_001074239.1 | 3 | 287 | ||
| NM_001080772.1 | NP_001074241.1 | 3 | 287 | |||
| NM_002255.5 | NP_002246.5 | 3 | 287 | |||
| GenBank mRNA | AF034771.1 | - | 3 | 253 | ||
| AF034772.1 | - | 3 | 253 | |||
| AF034773.1 | - | 3 | 253 | |||
| AF276292.1 | - | 3 | 247 | |||
| AF285436.1 | - | 3 | 274 | |||
| AK309531.1 | - | 4 | 523 | |||
| AK310066.1 | - | 3 | 529 | |||
| AY223512.2 | - | 1 | 174 | |||
| AY223513.1 | - | 1 | 174 | |||
| AY223514.1 | - | 1 | 174 | |||
| AY223515.1 | - | 1 | 174 | |||
| AY250088.1 | - | 1 | 174 | |||
| AY359817.1 | - | 3 | 258 | |||
| AY601812.1 | - | 3 | 241 | |||
| AY789058.1 | - | 3 | 241 | |||
| AY789059.1 | - | 3 | 241 | |||
| AY789060.1 | - | 3 | 241 | |||
| AY789061.1 | - | 3 | 241 | |||
| AY789062.1 | - | 3 | 241 | |||
| BC041611.1 | - | 3 | 288 | 5485491 | ||
| BM918119.1 | - | 287 | ||||
| DA934665.1 | - | 3 | 529 | |||
| DQ266438.1 | - | 3 | 250 | |||
| EU194340.1 | - | 3 | 247 | |||
| EU194342.1 | - | 3 | 247 | |||
| X97229.1 | - | 3 | 252 | |||
| X99479.1 | - | 3 | 247 | |||
| X99480.1 | - | 3 | 258 | |||
| Y13054.1 | - | 3 | 247 | |||

KIR2DL4
Genomic Map