siRNA Details
| Gene Symbol: THEMIS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010923.2 | NP_001010923.1 | 1, 2 | 237 | ||
| NM_001164685.1 | NP_001158157.1 | 1, 2 | 237 | |||
| XM_011535813.2 | XP_011534115.1 | 2, 3 | 404 | |||
| XM_011535816.1 | XP_011534118.1 | 1, 2 | 204 | |||
| XM_017010848.1 | XP_016866337.1 | 2, 3 | 404 | |||
| XM_017010849.1 | XP_016866338.1 | 2, 3 | 404 | |||
| GenBank mRNA | AK097903.1 | - | 1, 2 | 214 | ||
| AK292046.1 | - | 1, 2 | 187 | |||
| BC130516.1 | - | 1, 2 | 231 | 40146547 | ||
| BC144034.1 | - | 1, 2 | 229 | |||
| DA818250.1 | - | 2, 3 | 405 | |||

THEMIS
Genomic Map