siRNA Details
| Gene Symbol: RTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134888.2 | NP_001128360.1 | 1 | 922 | ||
| GenBank mRNA | BC150617.1 | - | 1 | 922 | 9056987 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RTL1| Gene Symbol: RTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134888.2 | NP_001128360.1 | 1 | 922 | ||
| GenBank mRNA | BC150617.1 | - | 1 | 922 | 9056987 | |