siRNA Details
| Gene Symbol: FAM132A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014980.2 | NP_001014980.1 | 2 | 238 | ||
| XM_011541435.2 | XP_011539737.1 | 2 | 336 | |||
| XM_011541436.2 | XP_011539738.1 | 2 | 336 | |||
| XM_011541437.1 | XP_011539739.1 | 2 | 80 | |||
| GenBank mRNA | BC089443.1 | - | 2 | 238 | 30554756 | |

FAM132A
Genomic Map