siRNA Details
| Gene Symbol: TMEM82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013641.2 | NP_001013663.1 | 5 | 995 | ||
| GenBank mRNA | BC127706.1 | - | 5 | 975 | 40127826 | |
| BC137239.1 | - | 5 | 995 | 9021236 | ||
| BC137240.1 | - | 5 | 995 | 9021237 | ||

TMEM82
Genomic Map