siRNA Details
Gene Symbol: CD164L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY358761.1 | - | 2 | 267 | ||
BC137466.1 | - | 2 | 256 | 9021466 | ||
DB114521.1 | - | 2 | 298 | |||
DB231017.1 | - | 2 | 279 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CD164L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY358761.1 | - | 2 | 267 | ||
BC137466.1 | - | 2 | 256 | 9021466 | ||
DB114521.1 | - | 2 | 298 | |||
DB231017.1 | - | 2 | 279 |