siRNA Details
| Gene Symbol: CCDC188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243537.1 | NP_001230466.1 | 6 | 578 | ||
| XM_005261238.3 | XP_005261295.1 | 8 | 1973 | |||
| XM_005261239.3 | XP_005261296.1 | 7 | 1917 | |||
| XM_005261240.3 | XP_005261297.1 | 9 | 2022 | |||
| XM_005261241.3 | XP_005261298.1 | 7 | 1865 | |||
| XM_011530171.2 | XP_011528473.1 | 7 | 1868 | |||
| GenBank mRNA | DY655741.1 | - | 4 | 343 | ||
| DY655742.1 | - | 1 | 18 | |||

CCDC188
Genomic Map