siRNA Details
| Gene Symbol: CCDC188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243537.1 | NP_001230466.1 | 4, 5 | 368 | ||
| XM_005261240.3 | XP_005261297.1 | 7, 8 | 1812 | |||
| XM_011530170.2 | XP_011528472.1 | 7, 8 | 1813 | |||
| GenBank mRNA | BU579471.1 | - | 4, 5 | 370 | ||
| DY655741.1 | - | 2, 3 | 133 | |||

CCDC188
Genomic Map