siRNA Details
| Gene Symbol: LRRC75B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207644.2 | NP_997527.2 | 3 | 368 | ||
| XM_005261600.2 | XP_005261657.1 | 4 | 642 | |||
| GenBank mRNA | AK097054.1 | - | 3 | 2084 | ||
| BC017595.1 | - | 2 | 616 | |||
| BC032388.1 | - | 3 | 329 | |||
| BC108684.1 | - | 3 | 370 | 6471196 | ||
| BI544432.1 | - | 3 | 380 | |||

LRRC75B
Genomic Map