siRNA Details
| Gene Symbol: LRRC75B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207644.2 | NP_997527.2 | 4 | 737 | ||
| GenBank mRNA | AK097054.1 | - | 4 | 2453 | ||
| BC017595.1 | - | 3 | 985 | |||
| BC032388.1 | - | 4 | 698 | |||
| BC108684.1 | - | 4 | 739 | 6471196 | ||

LRRC75B
Genomic Map