siRNA Details
| Gene Symbol: MUSTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_205853.3 | NP_995325.3 | 2 | 355 | ||
| GenBank mRNA | AJ276556.1 | - | 2 | 200 | ||
| BC130550.1 | - | 2 | 126 | 40146581 | ||
| BC130552.1 | - | 2 | 124 | 40146583 | ||
| BX089772.1 | - | 2 | 340 | |||
| CB043913.1 | - | 2 | 365 | |||
| CR990075.1 | - | 2 | 246 | |||
| Gene Symbol: TMEM110-MUSTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198974.2 | NP_001185903.2 | 9 | 1085 | ||
| GenBank mRNA | AL833659.1 | - | 3 | 4894 | ||

MUSTN1
Genomic Map