siRNA Details
| Gene Symbol: ANKUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144960.2 | NP_001138432.1 | 5 | 1253 | ||
| NM_001315505.1 | NP_001302434.1 | 5 | 1143 | |||
| NM_001315506.1 | NP_001302435.1 | 5 | 1253 | |||
| XM_011512798.1 | XP_011511100.1 | 5 | 750 | |||
| XM_011512799.1 | XP_011511101.1 | 5 | 984 | |||
| XM_011512801.1 | XP_011511103.1 | 3 | 421 | |||
| GenBank mRNA | AK027233.1 | - | 2 | 252 | ||
| AK304359.1 | - | 5 | 984 | |||
| BC171805.1 | - | 5 | 1143 | |||

ANKUB1
Genomic Map