siRNA Details
| Gene Symbol: EGFEM1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021485.2 | 2, 3 | 651 | |||
| GenBank mRNA | BI461452.1 | - | 2, 3 | 514 | ||
| DR000794.1 | - | 2, 3 | 658 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EGFEM1P| Gene Symbol: EGFEM1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021485.2 | 2, 3 | 651 | |||
| GenBank mRNA | BI461452.1 | - | 2, 3 | 514 | ||
| DR000794.1 | - | 2, 3 | 658 | |||