siRNA Details
| Gene Symbol: TMPRSS11F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207407.2 | NP_997290.2 | 8 | 1006 | ||
| GenBank mRNA | AK122625.1 | - | 8 | 1006 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS11F| Gene Symbol: TMPRSS11F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207407.2 | NP_997290.2 | 8 | 1006 | ||
| GenBank mRNA | AK122625.1 | - | 8 | 1006 | ||