siRNA Details
| Gene Symbol: LINC00615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038868.1 | 5 | 498 | |||
| GenBank mRNA | AK058107.1 | - | 5 | 497 | ||
| Gene Symbol: LOC105374972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134614.1 | 3 | 344 | |||
| GenBank mRNA | BX113657.1 | - | 3 | 344 | ||

LINC00615
Genomic Map