siRNA Details
| Gene Symbol: C6orf222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010903.4 | NP_001010903.3 | 12 | 2123 | ||
| XM_011514596.2 | XP_011512898.1 | 12 | 2976 | |||
| XM_011514597.2 | XP_011512899.1 | 12 | 2250 | |||
| XM_011514598.2 | XP_011512900.1 | 11 | 1633 | |||
| GenBank mRNA | BC140873.1 | - | 12 | 2046 | 9021741 | |
| BX648558.1 | - | 12 | 2124 | |||

C6orf222
Genomic Map