siRNA Details
| Gene Symbol: MROH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207414.2 | NP_997297.2 | 12, 13 | 1531 | ||
| GenBank mRNA | AK125848.1 | - | 12, 13 | 1531 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MROH5| Gene Symbol: MROH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207414.2 | NP_997297.2 | 12, 13 | 1531 | ||
| GenBank mRNA | AK125848.1 | - | 12, 13 | 1531 | ||