siRNA Details
| Gene Symbol: CFAP77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207417.2 | NP_997300.1 | 2 | 325 | ||
| GenBank mRNA | AK128819.1 | - | 2 | 297 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFAP77| Gene Symbol: CFAP77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207417.2 | NP_997300.1 | 2 | 325 | ||
| GenBank mRNA | AK128819.1 | - | 2 | 297 | ||