siRNA Details
| Gene Symbol: OR52H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005289.1 | NP_001005289.1 | 1 | 60 | ||
| GenBank mRNA | BC136998.1 | - | 1 | 86 | 9020990 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52H1| Gene Symbol: OR52H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005289.1 | NP_001005289.1 | 1 | 60 | ||
| GenBank mRNA | BC136998.1 | - | 1 | 86 | 9020990 | |