siRNA Details
| Gene Symbol: OR52N2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005174.1 | NP_001005174.1 | 1 | 815 | ||
| XM_011520090.2 | XP_011518392.1 | 2 | 6491 | |||
| GenBank mRNA | BC140732.1 | - | 1 | 837 | 9021600 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52N2| Gene Symbol: OR52N2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005174.1 | NP_001005174.1 | 1 | 815 | ||
| XM_011520090.2 | XP_011518392.1 | 2 | 6491 | |||
| GenBank mRNA | BC140732.1 | - | 1 | 837 | 9021600 | |