siRNA Details
| Gene Symbol: OR11H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004479.1 | NP_001004479.1 | 1 | 183 | ||
| GenBank mRNA | BC137054.1 | - | 1 | 236 | 9021051 | |
| BC137055.1 | - | 1 | 236 | 9021052 | ||
More Information
Set Membership: |
|

OR11H4
Genomic Map