siRNA Details
| Gene Symbol: RNASE9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001673.3 | NP_001001673.2 | 2 | 565 | ||
| NM_001110356.1 | NP_001103826.1 | 3 | 617 | |||
| NM_001110357.1 | NP_001103827.1 | 3 | 600 | |||
| NM_001110358.1 | NP_001103828.1 | 4 | 653 | |||
| NM_001110359.1 | NP_001103829.1 | 5 | 763 | |||
| NM_001110360.1 | NP_001103830.1 | 4 | 728 | |||
| NM_001110361.1 | NP_001103831.1 | 5 | 688 | |||
| NM_001289110.1 | NP_001276039.1 | 4 | 652 | |||
| GenBank mRNA | AF382949.1 | - | 1 | 493 | ||
| AY665804.1 | - | 1 | 493 | |||
| AY907670.1 | - | 2 | 565 | |||
| BC130311.1 | - | 1 | 578 | 40146342 | ||
| BC130313.1 | - | 1 | 578 | 40146344 | ||
| EF061296.1 | - | 3 | 600 | |||
| EF061297.1 | - | 3 | 617 | |||
| EF061298.1 | - | 4 | 652 | |||
| EF061299.1 | - | 4 | 653 | |||
| EF061300.1 | - | 4 | 728 | |||
| EF061301.1 | - | 5 | 688 | |||
| EF061302.1 | - | 5 | 763 | |||
| EF061303.1 | - | 3 | 2184 | |||
| EF061304.1 | - | 4 | 2219 | |||
| EU414264.1 | - | 1 | 493 | |||

RNASE9
Genomic Map