siRNA Details
| Gene Symbol: GDPGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013657.2 | NP_001013679.2 | 4 | 643 | ||
| NM_001322811.1 | NP_001309740.1 | 5 | 762 | |||
| GenBank mRNA | AK130527.1 | - | 4 | 1229 | ||
| BC127821.1 | - | 4 | 672 | |||

GDPGP1
Genomic Map