siRNA Details
| Gene Symbol: OR2AK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004491.1 | NP_001004491.1 | 1 | 705 | ||
| GenBank mRNA | BC136816.1 | - | 1 | 804 | 9020807 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR2AK2| Gene Symbol: OR2AK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004491.1 | NP_001004491.1 | 1 | 705 | ||
| GenBank mRNA | BC136816.1 | - | 1 | 804 | 9020807 | |