siRNA Details
Gene Symbol: TTC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242672.1 | NP_001229601.1 | 5 | 1009 | ||
XM_016999990.1 | XP_016855479.1 | 5 | 1081 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TTC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242672.1 | NP_001229601.1 | 5 | 1009 | ||
XM_016999990.1 | XP_016855479.1 | 5 | 1081 |