siRNA Details
Gene Symbol: HMCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291815.1 | NP_001278744.1 | 22 | 3253 | ||
XM_011518465.2 | XP_011516767.1 | 22 | 3390 | |||
XM_011518466.2 | XP_011516768.1 | 22 | 3390 | |||
XM_011518467.2 | XP_011516769.1 | 22 | 3390 | |||
XM_011518468.2 | XP_011516770.1 | 22 | 3390 | |||
XM_011518469.2 | XP_011516771.1 | 22 | 3390 | |||
XM_011518470.2 | XP_011516772.1 | 22 | 3390 | |||
GenBank mRNA | AK128020.1 | - | 10 | 1213 | ||
BC146865.1 | - | 9 | 1101 | 9056793 |
