siRNA Details
| Gene Symbol: LCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005565.3 | NP_005556.1 | 3, 4 | 398 | ||
| GenBank mRNA | AK292890.1 | - | 3, 4 | 359 | ||
| AK300000.1 | - | 3, 4 | 377 | |||
| AK310369.1 | - | 3, 4 | 339 | |||
| BC016618.1 | - | 3, 4 | 335 | 4476274 | ||
| BT007273.1 | - | 3, 4 | 191 | |||
| FN432830.1 | - | 3, 4 | 191 | |||
| U20158.1 | - | 3, 4 | 446 | |||

LCP2
Genomic Map