siRNA Details
| Gene Symbol: LIMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204426.1 | NP_001191355.1 | 5, 6 | 709 | ||
| NM_002314.3 | NP_002305.1 | 6, 7 | 931 | |||
| GenBank mRNA | AB209557.1 | - | 3, 4 | 403 | ||
| AF134379.1 | - | 6, 7 | 717 | |||
| AK290162.1 | - | 7, 8 | 856 | |||
| AK300382.1 | - | 5, 6 | 648 | |||
| D26309.1 | - | 7, 8 | 812 | |||

LIMK1
Genomic Map