siRNA Details
| Gene Symbol: LLGL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015002.1 | NP_001015002.1 | 6 | 574 | ||
| NM_001031803.1 | NP_001026973.1 | 6 | 574 | |||
| NM_004524.2 | NP_004515.2 | 6 | 574 | |||
| XM_011524801.1 | XP_011523103.1 | 6 | 557 | |||
| XM_011524802.1 | XP_011523104.1 | 6 | 545 | |||
| XM_011524807.2 | XP_011523109.1 | 6 | 662 | |||
| XM_017024625.1 | XP_016880114.1 | 6 | 545 | |||
| XM_017024626.1 | XP_016880115.1 | 6 | 545 | |||
| XM_017024627.1 | XP_016880116.1 | 6 | 545 | |||
| XM_017024628.1 | XP_016880117.1 | 6 | 545 | |||
| XM_017024629.1 | XP_016880118.1 | 6 | 545 | |||
| XM_017024630.1 | XP_016880119.1 | 6 | 545 | |||
| XM_017024631.1 | XP_016880120.1 | 6 | 545 | |||
| GenBank mRNA | AK126687.1 | - | 3 | 597 | ||
| AK315126.1 | - | 6 | 524 | |||
| BC006503.2 | - | 7 | 632 | |||
| BC010879.1 | - | 6 | 575 | |||
| BC064994.1 | - | 6 | 547 | 5575350 | ||
| X87342.1 | - | 6 | 550 | |||

LLGL2
Genomic Map