siRNA Details
| Gene Symbol: C10orf120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010912.2 | NP_001010912.1 | 1, 2 | 211 | ||
| GenBank mRNA | AL704711.1 | - | 1, 2 | 157 | ||
| BC065903.1 | - | 1, 2 | 215 | |||
| BC140922.1 | - | 1, 2 | 203 | 9056734 | ||

C10orf120
Genomic Map