siRNA Details
| Gene Symbol: C11orf88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100388.1 | NP_001093858.1 | 4, 5 | 408 | ||
| NM_207430.2 | NP_997313.2 | 5, 6 | 489 | |||
| GenBank mRNA | AI829856.1 | - | 2, 3 | 308 | ||
| AK128145.1 | - | 5, 6 | 489 | |||
| BC039505.2 | - | 4, 5 | 393 | |||

C11orf88
Genomic Map