siRNA Details
Gene Symbol: PATE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144874.1 | NP_001138346.1 | 2 | 167 | ||
GenBank mRNA | AK123042.1 | - | 2 | 167 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PATE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144874.1 | NP_001138346.1 | 2 | 167 | ||
GenBank mRNA | AK123042.1 | - | 2 | 167 |