siRNA Details
| Gene Symbol: SNX19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014758.2 | NP_055573.2 | 1 | 1415 | ||
| XM_005271546.4 | XP_005271603.1 | 1 | 1414 | |||
| XM_011542819.2 | XP_011541121.1 | 1 | 1414 | |||
| XM_011542820.2 | XP_011541122.1 | 1 | 1414 | |||
| XM_011542821.2 | XP_011541123.1 | 1 | 1415 | |||
| XM_011542822.2 | XP_011541124.1 | 1 | 1414 | |||
| XM_017017725.1 | XP_016873214.1 | 1 | 1390 | |||
| GenBank mRNA | CN307729.1 | - | 1 | 334 | ||

SNX19
Genomic Map