siRNA Details
| Gene Symbol: FLJ41278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033988.1 | 2 | 176 | |||
| GenBank mRNA | AK123272.1 | - | 2 | 177 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ41278| Gene Symbol: FLJ41278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033988.1 | 2 | 176 | |||
| GenBank mRNA | AK123272.1 | - | 2 | 177 | ||