siRNA Details
| Gene Symbol: FAM177B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324080.1 | NP_001311009.1 | 2 | 176 | ||
| NM_207468.2 | NP_997351.2 | 3 | 278 | |||
| NR_136691.1 | 3 | 278 | ||||
| XM_017001279.1 | XP_016856768.1 | 2 | 720 | |||
| XM_017001280.1 | XP_016856769.1 | 3 | 234 | |||
| XM_017001281.1 | XP_016856770.1 | 3 | 803 | |||
| XM_017001282.1 | XP_016856771.1 | 3 | 259 | |||
| XM_017001283.1 | XP_016856772.1 | 2 | 52 | |||
| XM_017001284.1 | XP_016856773.1 | 2 | 52 | |||
| XM_017001285.1 | XP_016856774.1 | 2 | 52 | |||
| GenBank mRNA | AK125494.1 | - | 3 | 278 | ||

FAM177B
Genomic Map