siRNA Details
| Gene Symbol: C2orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213608.2 | NP_998773.1 | 1 | 955 | ||
| GenBank mRNA | AY358249.1 | - | 1 | 955 | ||
| BC137150.1 | - | 1 | 831 | 9021147 | ||
| BC137151.1 | - | 1 | 831 | 9021148 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2orf66| Gene Symbol: C2orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213608.2 | NP_998773.1 | 1 | 955 | ||
| GenBank mRNA | AY358249.1 | - | 1 | 955 | ||
| BC137150.1 | - | 1 | 831 | 9021147 | ||
| BC137151.1 | - | 1 | 831 | 9021148 | ||