siRNA Details
| Gene Symbol: TMPRSS11BNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123556.1 | - | 4 | 471 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS11BNL| Gene Symbol: TMPRSS11BNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK123556.1 | - | 4 | 471 | ||