siRNA Details
| Gene Symbol: PRR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214711.3 | NP_999876.2 | 3 | 593 | ||
| GenBank mRNA | BX640892.1 | - | 3 | 506 | ||
| BX640924.1 | - | 3 | 544 | |||
| BX640997.1 | - | 3 | 580 | |||
| BX647185.1 | - | 4 | 583 | |||
| BX648724.1 | - | 3 | 542 | |||
| BX648872.1 | - | 3 | 465 | |||
| BX648895.1 | - | 3 | 542 | |||
| BX648896.1 | - | 4 | 477 | |||

PRR27
Genomic Map