siRNA Details
| Gene Symbol: CRIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206922.2 | NP_996805.2 | 2, 3 | 144 | ||
| XM_005249103.3 | XP_005249160.1 | 2, 3 | 2057 | |||
| XM_011514609.2 | XP_011512911.1 | 2, 3 | 2057 | |||
| XM_017010856.1 | XP_016866345.1 | 2, 3 | 2057 | |||
| XM_017010857.1 | XP_016866346.1 | 2, 3 | 2057 | |||
| GenBank mRNA | AY555741.1 | - | 2, 3 | 144 | ||
| AY555742.1 | - | 2, 3 | 144 | |||
| AY555743.1 | - | 2, 3 | 144 | |||
| AY555744.1 | - | 2, 3 | 144 | |||
| CN282994.1 | - | 1, 2 | 91 | |||

CRIP3
Genomic Map