siRNA Details
| Gene Symbol: SNX30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012994.1 | NP_001013012.1 | 2 | 376 | ||
| XM_011518691.2 | XP_011516993.1 | 2 | 413 | |||
| XM_017014716.1 | XP_016870205.1 | 2 | 444 | |||
| GenBank mRNA | AK127013.1 | - | 1 | 80 | ||
| BG699098.1 | - | 2 | 397 | |||
| BX367137.2 | - | 2 | 375 | |||

SNX30
Genomic Map