siRNA Details
| Gene Symbol: FIGNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013690.4 | NP_001013712.4 | 2 | 1639 | ||
| XM_005268870.3 | XP_005268927.1 | 2 | 1692 | |||
| XM_017019298.1 | XP_016874787.1 | 2 | 1881 | |||
| XM_017019299.1 | XP_016874788.1 | 3 | 2437 | |||

FIGNL2
Genomic Map