siRNA Details
| Gene Symbol: C6orf201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085401.2 | NP_001078870.1 | 4 | 1123 | ||
| NR_104463.1 | 5 | 1380 | ||||
| NR_104464.1 | 3 | 745 | ||||
| XM_011514613.2 | XP_011512915.1 | 4 | 737 | |||
| XM_017010861.1 | XP_016866350.1 | 3 | 754 | |||
| GenBank mRNA | AK124114.1 | - | 4 | 815 | ||
| AK301893.1 | - | 3 | 558 | |||
| BC066986.1 | - | 3 | 734 | 4827732 | ||

C6orf201
Genomic Map