siRNA Details
| Gene Symbol: C6orf201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085401.2 | NP_001078870.1 | 2 | 806 | ||
| NR_104463.1 | 2 | 806 | ||||
| XM_005249112.3 | XP_005249169.1 | 1 | 283 | |||
| XM_011514613.2 | XP_011512915.1 | 2 | 420 | |||
| GenBank mRNA | AF429972.1 | - | 1 | 256 | ||
| AK124114.1 | - | 1 | 241 | |||
| AK301893.1 | - | 1 | 241 | |||
| BC047663.1 | - | 2 | 803 | 5266746 | ||

C6orf201
Genomic Map