siRNA Details
| Gene Symbol: ANKHD1-EIF4EBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020690.5 | NP_065741.3 | 34, 35 | 7825 | ||
| GenBank mRNA | AB029008.1 | - | 5, 6 | 4931 | ||
| AF521883.1 | - | 34, 35 | 7731 | |||
| AK125734.1 | - | 7, 8 | 1789 | |||
| BC144623.1 | - | 34, 35 | 7910 | |||

ANKHD1-EIF4EBP3
Genomic Map