siRNA Details
| Gene Symbol: MANBA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005908.3 | NP_005899.3 | 6 | 841 | ||
| XM_017008203.1 | XP_016863692.1 | 7 | 827 | |||
| XM_017008204.1 | XP_016863693.1 | 5 | 672 | |||
| GenBank mRNA | AB209846.1 | - | 7 | 866 | ||
| AK291598.1 | - | 6 | 829 | |||
| AK300015.1 | - | 5 | 611 | |||
| AK312827.1 | - | 6 | 796 | |||
| BC015743.1 | - | 6 | 831 | 4905535 | ||
| EU009130.1 | - | 6 | 741 | |||
| U60337.1 | - | 6 | 828 | |||

MANBA
Genomic Map