siRNA Details
| Gene Symbol: MAP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033063.1 | NP_149052.1 | 1 | 955 | ||
| NM_207577.1 | NP_997460.1 | 1 | 955 | |||
| XM_006718556.3 | XP_006718619.1 | 1 | 3594 | |||
| XM_011545039.2 | XP_011543341.1 | 1 | 3595 | |||
| XM_017017755.1 | XP_016873244.1 | 1 | 3494 | |||
| XM_017017757.1 | XP_016873246.1 | 1 | 48 | |||
| GenBank mRNA | AB058781.1 | - | 1 | 955 | ||
| BC063860.1 | - | 1 | 621 | |||
| BC139780.1 | - | 2 | 194 | 40122938 | ||
| BC150254.1 | - | 1 | 955 | 9007181 | ||

MAP6
Genomic Map