siRNA Details
| Gene Symbol: DNAJB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001394.3 | NP_001001394.1 | 1 | 434 | ||
| GenBank mRNA | AY083665.1 | - | 1 | 434 | ||
| BC017590.2 | - | 1 | 434 | 4828023 | ||
| BC024013.2 | - | 1 | 397 | 4826480 | ||
| Gene Symbol: LOC100286922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037694.1 | 1 | 411 | |||
| GenBank mRNA | BC066346.1 | - | 1 | 396 | ||

DNAJB3
Genomic Map