siRNA Details
Gene Symbol: MC5R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005913.2 | NP_005904.1 | 1 | 1059 | ||
GenBank mRNA | BC069153.1 | - | 1 | 838 | 7217013 | |
BC069545.1 | - | 1 | 837 | 7262232 | ||
BC095531.1 | - | 1 | 1062 | 6971958 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
