siRNA Details
Gene Symbol: ADAM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318933.1 | NP_001305862.1 | 7, 8 | 629 | ||
NM_002390.5 | NP_002381.2 | 7, 8 | 629 | |||
XM_005257373.3 | XP_005257430.1 | 7, 8 | 771 | |||
XM_017024647.1 | XP_016880136.1 | 6, 7 | 547 | |||
GenBank mRNA | AB009675.1 | - | 7, 8 | 608 | ||
AK296512.1 | - | 7, 8 | 710 | |||
CD630831.1 | - | 7, 8 | 580 | |||
D17390.1 | - | 4, 5 | 338 |
